Learn More
Cytiva VacuCap™ 60 Vacuum Filtration Devices
Remove endotoxin from water, buffer, neutral sugars, and certain biological solutions.
Brand:  Cytiva TA4632
432.25 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Product Code. 17134821
  • 455.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/4 in. hose barb inner taper
0.2 μm
10/Pk.
Hydrophilic PES
Sterile
60 mm