Learn More
Cytiva VacuCap™ 90 Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
Brand: Cytiva 4624
This item is not returnable.
View return policy
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Supor (Hydrophilic PES) | |
Sterile | |
90 mm |
0.45 μm | |
60 cm2 |