missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) VacuCap™ 90 Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
Brand: Cytiva (formerly Pall Lab) 4632
233.17 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Supor (Hydrophilic PES) | |
Sterile | |
60 mm |
0.2 μm | |
30 cm2 |