Learn More
Cytiva VacuCap™ 90 Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
Brand:  Cytiva 4634
 View more versions of this product
Product Code. 17184371
  • 219.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Supor (Hydrophilic PES)
Sterile
60 mm
0.45 μm
30 cm2