Learn More
Greiner Bio-One Vacuette Tube™, Premium, Red Cap-Yellow Ring, CAT Serum Separator Clot Activator, Sterile
Includes inner wall coated with microscopic silica particles to activate coagulation and a separator gel to separate serum and blood cells.
Brand:  Greiner Bio-One 454028
Product Code. 10060704
  • 36.65 € / Pack of 50
Explore more special offers
Description
Description
  • The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.
  • Serum separator tubes contain a separator gel in the base of the tube; during centrifugation, this gel forms a stable barrier between the serum and the blood cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuette Tube
Sterile
Red with Yellow Ring
13 mm
CAT Serum Sep Clot Activator/Gel Separator
2.5 mL
75 mm