missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Glass ACD Solution Tube with Yellow Closure
Used to obtain a whole blood or a plasma sample. BD Vacutainer™ Glass ACD Solution Tube with Yellow Closure contains the active anticoagulant Tri-sodium Citrate with Citric Acid and Dextrose. Two formulations of the ACD solution are available.
Brand: BD 367756
This item is not returnable.
View return policy
Description
BD Vacutainer™ ACD tubes are available in glass only and are identifiable by a yellow cap (Hemogard™ or conventional). They contain the active anticoagulant Tri-sodium Citrate with Citric Acid and Dextrose in one of two formulations. The concentration of additives in each solution is as follows:- ACD Solution A (conventional closure)
- Sodium citrate: 22.0g/L
- Dextrose: 24.5g/L
- Citric acid: 8.0g/L
- Antimycotic (K sorbate) reagent: 0.15g/L
- ACD Solution B (Hemogard™ closure)
- Sodium citrate: 13.2g/L
- Dextrose: 14.7g/L
- Citric acid: 4.8g/L
- Antimycotic (K sorbate) reagent: 0.2g/L

Specifications
ACD Solution B Tube | |
Glass | |
Hemogard | |
Yellow | |
Used to obtain a whole blood or plasma sample |
ACD Solution B | |
Sterile | |
6 mL | |
13 x 100 mm | |
Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction