missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube

Product Code. p-8000826
Click to view available options
Capacity (Metric):
6 mL
10 mL
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12957666

Brand: BD 367873

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used to obtain a whole blood or EDTA plasma sample. Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube contains K2EDTA which is a liquid solution that is spray coated onto the interior surface of the BD Vacutainer Plus Tubes.

Lavender Hemogard™ closure
Paper label
TRUSTED_SUSTAINABILITY

Specifications

Product Type K2 EDTA Tube
Additive K2 EDTA
Material Plastic
Closure Type Hemogard
Capacity (Metric) 6 mL
Closure Color Lavender
Height (Metric) 100 mm
Label Type Block
Diameter (Metric) Outer 13 mm
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.