missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Hemogard™ Closure Plastic K3-Edta Tube

Use with whole blood or EDTA plasma sample. BD Vacutainer™ Hemogard™ Closure Plastic K3-EDTA Tube contains the additive K3 Potassium Salt of EDTA that acts as a coagulant.

Brand:  BD 368860


 View more versions of this product

Product Code. 12987666

  • 138.00 € / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lavender Hemogard™ Closure
  • Used for whole blood or EDTA plasma sample
  • The additive K3 Potassium Salt of EDTA is a liquid in the Vacutainer Tube and acts as a coagulant
  • Available in sizes 2ml, 3ml and 4ml.
  • The difference between K2EDTA and K3EDTA is that the K3 is a liquid solution in the tubes, whereas the K2 is a liquid solution that is spray coated onto the tubes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

K2 EDTA Tube
Plastic
4 mL
75 mm
Block
K3 EDTA Additive
Hemogard
Lavender
Used to obtain a whole blood or EDTA plasma sample
13 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X1000 BD Vacutainer 13 x 75 mm x 4.0ml EDTA

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H332-Harmful if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Tripotassium EDTA Dihydrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.