missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Plastic K2EDTA Tube with Lavender Hemogard™ closure

Product Code. p-8009459
Click to view available options
Capacity (Metric):
3 mL
4 mL
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 12947666

Brand: BD 367839

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with whole blood or EDTA plasma sample. BD Vacutainer™ Plastic K2EDTA Tube with Lavender Hemogard™ closure includes the coagulant K2 which is a liquid solution that is spray coated onto the tubes.

EDTA is spray coated onto the interior surface of the Vacutainer Tube
  • It acts as a coagulant
  • Available in sizes 3ml, 4ml, 6ml and 10ml
  • The difference between K2EDTA and K3EDTA is that the K3 is a liquid solution in the tubes, whereas the K2 is a liquid solution that is spray coated onto the tubes
  • Lavender Hemogard™ closure
  • TRUSTED_SUSTAINABILITY

    Specifications

    Material Plastic
    Capacity (Metric) 4 mL
    compliance-icons
    Product Identifier
    • X1000 EDTA tube BD Vacutainer K2EDTA tube with BD
    Signal Word
    • Warning
    Hazard Category
    • Acute toxicity Category 4
    Hazard Statement
    • H332-Harmful if inhaled.
    Precautionary Statement
    • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
    • P271-Use only outdoors or in a well-ventilated area.
    • P312-Call a POISON CENTER/doctor/if you feel unwell.
    Supplemental information
    • MIXTURE LIST-Contains : Dipotassium EDTA Dihydrate
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.