Learn More
BD Vacutainer™ Plastic Urinalysis Tubes
For the collection and transport of urine specimens. BD Vacutainer™ Plastic Urinalysis Tubes are sterile evacuated tubes with or without preservative to be used with BD collection devices. Available in many sizes with a round or conical bottom.
Brand:  BD 368500
 View more versions of this product
Product Code. 12937766
  • 128.00 € / Pack of 1000
Explore more special offers
Description
Description
BD Vacutainer™ urine products for the collection and transport of urine specimens offer the same level of safety and standardization as BD blood collection products.
  • Designed to provide accurate patient results with reduced risk of healthcare worker exposure to hazardous specimens
  • Used with BD collection devices, specimen cups, 24 hour 3L containers and transfer straws
  • Leak-proof urine tubes can safely transport urine specimens to the laboratory for analysis
  • Options include bottom shape (round or conical) and cap style (conventional or Hemogard™)
  • Most sizes are preservative/additive free
  • 8mL tube (mfr. no. 364938) contains a preservative: 0.4% Chlorhexidine, 5.6% Ethyl Paraben and 94% Sodium Propionate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Urinalysis Tube
Hemogard™
PET
No additive
Urinalysis
Paper
4 mL
Beige
Single-use
Sterile
Round
Urine collection devices (cups, transfer straws, 24 hr. containers)
100 Pk.