Learn More
BD Vacutainer™ Push Button Blood Collection Sets
Brand:  BD 367355
 View more versions of this product
Product Code. 12947746
  • 168.00 € / Pack of 100
Explore more special offers
Specifications
Push Button Blood Collection Set
21 G
19 mm
100 Cs.
Green
21 guage, 19mm needle size, 178mm tubing
178 mm