missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Serum Tubes

Used to obtain a serum sample. BD™ Vacutainer™ Serum Tubes are made of durable PET plastic with a Hemogard™ closure. It has silica particles added to trigger coagulation, and are marked CAT and CE.

Brand:  BD 368975


 View more versions of this product

Product Code. 12937686

  • 145.00 € / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

In order to obtain serum samples from plastic tubes, the tube must have a coagulation activator added. As the plastic surface alone is insufficient to trigger the coagulation within an acceptable time, BD Vacutainer™ plastic serum tubes have silica particles added for this purpose.

  • These tubes are marked with the acronym CAT (Clot Activator Tube)
  • Recommended minimum time for coagulation from patients who have not been treated with anticoagulants is 60 minutes
  • Centrifuging conditions: ≤1300 g for 10 minutes at 18–25°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serum Tube
PET
4 mL
13 x 75 mm
Block
Silica
Hemogard
Red
Used to obtain a serum sample
CE, CAT
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TUBE PLN PLH 13X75 4.0 BLBLCE RD

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A

Hazard Statement
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Quatrz

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.