Learn More
Welch Ilmvac™ Vacuum Filtration And Desiccation Connection Kit
Facilitates proper connection to vacuum filtration or dissicator. Welch Ilmvac™ Vacuum Filtration And Desiccation Connection Kit includes a filter and suitable tubing.
Brand: Welch Ilmvac™ 404008
This item is not returnable.
View return policy
Description
- Quick connection to vacuum filtration funnel / manifold or dessicator
- Kit includes in-line hydrophobic filter, 1.5m silicone vacuum hose (6mm I.D.) and 1.5m silicone vacuum hose (8mm I.D.)
Specifications
In-Line Hydrophobic Filter, 1.5m Silicone Vacuum Hose (6mm ID) And 1.5m Silicone Vacuum Hose (8mm ID) | |
1 Pack | |
Vacuum Filtration And Desiccation Connection Kit |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Fisherbrand Vacuum pumps
File size: 5.6 MB
Product Content Correction
Product Title
Welch Ilmvac™ Vacuum Filtration And Desiccation Connection Kit > Includes: In-Line Hydrophobic Filter, 1.5m Silicon
Spot an opportunity for improvement?Share a Content Correction