missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Vacuum Oven Thermoshelf
600.00 € - 810.00 €
Specifications
Certifications/Compliance | Calibration certificate |
---|---|
Type | Shelf |
For Use With (Application) | Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control) |
Includes | Local Temperature Sensing (Pt100, 4-Wire-Circuit) |
Product Code | Brand | For Use With (Equipment) | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Material | Price | Quantity & Availability | |||||
11741464
|
Memmert™
VO29B00741 |
Model 200 | Aluminum |
600.00 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
11751464
|
Memmert™
VO49B00743 |
Model 400 | Aluminum |
692.00 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
11991449
|
Memmert™
VO29B00733 |
Model 200 | Stainless Steel |
600.00 €
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
11323692
|
Memmert™
VO101B00744 |
Model 500 | Aluminum |
810.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
11901459
|
Memmert™
VO49B00734 |
Model 400 | Stainless Steel |
692.00 €
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
11911459
|
Memmert™
VO101B00735 |
Model 500 | Stainless Steel |
810.00 €
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||||
Specifications
Calibration certificate | |
Shelf | |
Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control) | |
Local Temperature Sensing (Pt100, 4-Wire-Circuit) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Vacuum Oven Thermoshelf