Learn More
Heidolph™ Vacuum Pump Hei-VAC Vario Control
Hei-VAC Vario Control is an RPM-regulated vacuum pump.
Brand:  Heidolph™ 591-00141-03
4316.54 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 17333160
  • 4555.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For Hei-VAP Expert Control / Hei-VAP Ultimate Control
  • Even if the gas ballast valve is open an excellent ultimate vacuum is reached when working with easily condensable vapors
  • This makes distilling high boiling point solvents such as DMF or DMSO possible at low bath temperatures
  • Condenser is not in scope of delivery
Specifications
Specifications
UK Plug
IP20
230 V