Learn More
Heidolph™ Vacuum Pump Hei-VAC Vario Station
Hei-VAC Vario Station is an RPM-regulated vacuum pump.
Brand: Heidolph™ 591-00142-03
This item is not returnable.
View return policy
Description
- Fully controllable stand-alone pumping unit including vacuum controller
- For Hei-VAP Core models
- Precise vacuum control avoids bumping and eliminates foaming of your evaporation solution
- Due to automatic vacuum supply to the process parameters you have more time for important laboratory activities
- Condenser is not in scope of delivery
Specifications
UK Plug | |
IP20 | |
230 V |