Learn More
Welch Ilmvac™ Vacuum Pump Mpc 110E Diaphragm Chemically Resistant
Vacuum Pump, Mpc 110E Diaphragm Chemically Resistant, 90-260V 50/60Hz 235mm x 140mm x 277mm 20L/min flow rate, <50 mbar ultimate vacuum
Brand:  Welch Ilmvac™ 412421-02
Product Code. 12602623
  • 1275.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Vacuum Pump, Mpc 110E Diaphragm Chemically Resistant, 90-260V 50/60Hz 235mm x 140mm x 277mm 20L/min flow rate, <50 mbar ultimate vacuum
90-260V 50/60Hz 235mm x 140mm x 277mm 20L/min flow rate, <50 mbar ultimate vacuum
Specifications
Specifications
50 Mbar
20 L/min
Vacuum Pump