Learn More
Welch Ilmvac Vacuum pump P 4 E rotary vane 230V 50/60Hz 405mm x 150mm x 215mm (w x d x h) pumping speed 77/92L/min ultimate vacuum 1 x 10^-1 mbar Welch-Ilmvac
Marca: Welch Ilmvac 321002
Vacuum pump P 4 E rotary vane 230V 50/60Hz 405mm x 150mm x 215mm (w x d x h) pumping speed 77/92L/min ultimate vacuum 1 x 10^-1 mbar Welch-Ilmvac
Code : 50
Detalles adicionales : Weight : 17.00000kg