missing translation for 'onlineSavingsMsg'
Learn More

VACUUBRAND™ VacuuSelect™ Touch Screen Vacuum Controllers
GREENER_CHOICE

Intuitive operation of rotary evaporation, short path distillation and more

Brand:  VACUUBRAND™ 20700100

 View more versions of this product

Product Code. 15922861

  • 3370.00 € / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Automatically finds boiling points
  • Pre-programmed routines for vacuum ovens, gel dryers, filtration and more
  • Assemble custom routines in minutes
  • For rotary evaporators and other diaphragm pump applications
  • Pirani control packages for short path distillation and other rotary vane/scroll pump applications
  • Two different Pirani packages for pumps with KF16 or KF25 flanges
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 to 3 mbar/Torr/hPa
100 to 240V, 50 to 60 Hz, universal
CEE/US/CH/UK/AUS/CN
Molecular Distillation
100 to 240 VAC
Controller, solenoid valve, T-connector, appropriate KF16 clamping and centering rings, power supply
Alumina ceramic capacitive and Pirani transducers
Works with rotary vane pumps
1080 mbar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VACUUBRAND™ VacuuSelect™ Touch Screen Vacuum Controllers > Max vacuum: 10 to 3 mbar/Torr/hPa; Includes: Controller, solenoid valve, T-connector, appropriate KF16 clamping and centering rings, power supply

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.