Learn More
Thermo Scientific™ High Pressure Stainless Steel Nuts and Ferrules
Maintain the performance of your Thermo Scientific™ Valco™ Injector for HPLC with these syringe ports, ferrules, and nuts.
19.79 € - 87.49 €
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10639865
|
Thermo Scientific™
F-190 |
Two-piece Fingerlok 10-32 nut with PEEK ferrule; 1/16 in. stainless steel | 1 Each | N/A | |||||
10423305
|
Thermo Scientific™
U-320X |
Valco 10-32 male hex nut; Stainless steel; | 10 Pack | N/A | |||||
10446934
|
Thermo Scientific™
U-400X |
316 Stainless Steel Male Nut for 1/16” tubing (0.0625in) | 10 Pack |
87.49 €
Pack of 10 |
Estimated Shipment: 09-01-2025
Log in to see stock. |
||||
10170983
|
Thermo Scientific™
U-401X |
Stainless Steel Ferrule for 1/16” tubing (0.0625in) | 10 Pack | N/A | |||||
Description
Maintain the performance of your Thermo Scientific™ Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
- Designed for 10-32 port configurations
- Burr and contaminant free
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.