Learn More
Valeraldehyde, 97%, Thermo Scientific Chemicals
23.42 € - 91.56 €
Chemical Identifiers
CAS | 110-62-3 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.13 |
MDL Number | MFCD00007026 |
InChI Key | HGBOYTHUEUWSSQ-UHFFFAOYSA-N |
Synonym | valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde |
PubChem CID | 8063 |
ChEBI | CHEBI:84069 |
IUPAC Name | pentanal |
SMILES | CCCCC=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10715041
|
Thermo Scientific Acros
149561000 |
100 mL | Glass bottle |
23.42 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10356550
|
Thermo Scientific Acros
149562500 |
250 mL | Glass bottle |
30.45 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666164
|
Thermo Scientific Acros
149560010 |
1 L | Glass bottle |
91.56 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-62-3 | |
86.13 | |
HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
8063 | |
pentanal |
C5H10O | |
MFCD00007026 | |
valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
CHEBI:84069 | |
CCCCC=O |
Specifications
110-62-3 | |
0.8070g/mL | |
12°C | |
C5H10O | |
1.3900 to 1.3950 | |
01, 676 | |
0.807 | |
Solubility in water: 14g/L (20°C). Other solubilities: soluble in alcohol, ether, propylene glycol and, many organic solvents | |
CCCCC=O | |
86.13 | |
CHEBI:84069 | |
86.13 | |
Valeraldehyde, 98% |
-92.0°C | |
103.0°C | |
96% min. (GC) | |
CH3(CH2)3CHO | |
MFCD00007026 | |
15, 10088 | |
valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
pentanal | |
8063 | |
0.6 mPa.s (20°C) | |
97% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-784-4
missing translation for 'rtecsNumber' : YV3600000
missing translation for 'tsca' : TSCA
- Valeraldehyde
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Valeraldehyde
RUO – Research Use Only