Learn More
Valeric acid, 99%, Thermo Scientific Chemicals
24.30 € - 87.20 €
Chemical Identifiers
CAS | 109-52-4 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.133 |
MDL Number | MFCD00004413 |
InChI Key | NQPDZGIKBAWPEJ-UHFFFAOYSA-N |
Synonym | valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n |
PubChem CID | 7991 |
ChEBI | CHEBI:17418 |
IUPAC Name | pentanoic acid |
SMILES | CCCCC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483890
|
Thermo Scientific Alfa Aesar
A16238.AE |
100 mL |
24.30 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483347
|
Thermo Scientific Alfa Aesar
A16238.AP |
500 mL |
29.30 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493347
|
Thermo Scientific Alfa Aesar
A16238.0F |
2500 mL |
87.20 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Valeric acid, is used as a sex attractant of the sugar beet wireworm, Limonius californicus. It is used predominantly as an intermediate in the manufacture of flavors and perfumes, ester type lubricants, plasticizers and vinyl stabilizesrs. It is a food additive used as a synthetic flavoring substance dan adjuvant. It is an inert ingredient in controlling pest.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsValeric acid, is used as a sex attractant of the sugar beet wireworm, Limonius californicus. It is used predominantly as an intermediate in the manufacture of flavors and perfumes, ester type lubricants, plasticizers and vinyl stabilizesrs. It is a food additive used as a synthetic flavoring substance dan adjuvant. It is an inert ingredient in controlling pest.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
109-52-4 | |
102.133 | |
NQPDZGIKBAWPEJ-UHFFFAOYSA-N | |
7991 | |
pentanoic acid |
C5H10O2 | |
MFCD00004413 | |
valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n | |
CHEBI:17418 | |
CCCCC(=O)O |
Specifications
109-52-4 | |
184°C to 186°C | |
CH3(CH2)3CO2H | |
UN3265 | |
14,9904 | |
It is soluble in water. | |
CCCCC(=O)O | |
102.133 | |
CHEBI:17418 | |
99% | |
88°C (190°F) | |
1.408 |
-34°C | |
C5H10O2 | |
MFCD00004413 | |
969454 | |
valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n | |
NQPDZGIKBAWPEJ-UHFFFAOYSA-N | |
pentanoic acid | |
7991 | |
102.13 | |
0.938 | |
Unpleasant | |
Valeric acid |
Safety and Handling
GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 203-677-2
missing translation for 'rtecsNumber' : YV6100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only