missing translation for 'onlineSavingsMsg'
Learn More

Valerophenone, 99%, Thermo Scientific Chemicals

Product Code. 11468793
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Valerophenone is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valerophenone is used as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
 Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1009-14-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00009480
InChI Key XKGLSKVNOSHTAD-UHFFFAOYSA-N
Synonym valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
PubChem CID 66093
ChEBI CHEBI:36812
IUPAC Name 1-phenylpentan-1-one
SMILES CCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point °C to 9°C
Density 0.978 g/mL
Boiling Point 244°C to 245°C
Flash Point 102°C (215°F)
Assay Percent Range 99%
Refractive Index 1.5143
Quantity 25 g
Beilstein 1907717
Solubility Information Insoluble in water.
Formula Weight 162.23
Percent Purity 99%
Chemical Name or Material Valerophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'