missing translation for 'onlineSavingsMsg'
Learn More

Valeryl chloride, 98%, Thermo Scientific Chemicals

44.72 € - 273.36 €

Chemical Identifiers

CAS 638-29-9
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000757
InChI Key XGISHOFUAFNYQF-UHFFFAOYSA-N
Synonym valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
PubChem CID 61186
IUPAC Name pentanoyl chloride
SMILES CCCCC(Cl)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10194320
View Documents
Thermo Scientific Acros
169121000
100 g Glass bottle
44.72 €
100g
Please to purchase this item. Need a web account? Register with us today!
13408408
View Documents
Thermo Scientific Acros
169120010
1 kg Glass bottle
273.36 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

638-29-9
120.58
XGISHOFUAFNYQF-UHFFFAOYSA-N
61186
CCCCC(Cl)=O
C5H9ClO
MFCD00000757
valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
pentanoyl chloride

Specifications

638-29-9
Colorless to Yellow
125.0°C to 127.0°C
Authentic
C5H9ClO
1.4190 to 1.4210
02,301
valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
XGISHOFUAFNYQF-UHFFFAOYSA-N
pentanoyl chloride
61186
98%
Valeryl chloride, 98%
-110.0°C
0.9900g/mL
32°C
97.5% min. (GC)
CH3(CH2)3COCl
MFCD00000757
0.99
Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
CCCCC(Cl)=O
120.58
120.58
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 211-330-1

missing translation for 'tsca' : TSCA

Product Identifier
  • Valeryl chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.