Learn More
OHAUS™ Valor™ 2000 Scale
OHAUS™ Valor™ 2000 Scale offers large LED front and rear displays, making it ideal for use in dry or washdown environments.
Brand:  OHAUS™ 30072315
214.91 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 15496101
  • 318.00 € / 1 unit
Explore more special offers
Description
Description
  • The sturdy polycarbonate keypad offers protection from knives and other sharp instruments used in food preparation, processing and portioning applications.
  • A unique IP68 flow-thru design ensures that the NSF-certified Valor 2000 will drain moisture in the scale when used in wet conditions.
  • Stabilization time is less than a half-second.
  • Large front and rear LED displays allow for easy viewing so two operators can share one scale.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 Hours with Rechargeable Battery
6.00 kg
1.000000 g
V22PWE6T
1.000000 g
-10°C to +40°C
256 x 280 x 121 mm
ABS housing with stainless steel platform (PW), stainless steel housing and platform (XW), up-front level indicator
Dual LED
1.000000 g
IPX8
0.500 sec.
Gram, Kilogram, Ounce, Pound, Pound:Ounce
5.000 kg