missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valproic acid, sodium salt, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Histone deacetylase inhibitor (IC50 = 400 μM) that exhibits anticancer, anti-inflammatory and neuroprotective effects. Displays anticonvulsive activity via an increase in GABA levels and decreases Aα production in animal models of Alzheimer's disease. Also attenuates NMDA-mediated excitation, blocks voltage-gated Na+ channels and modulates firing of neurons. Enables induction of pluripotent stem cells from somatic cells by Oct4 and Sox2. Can induce autophagy by inhibiting inositol synthesis. Identified by chemoinformatics as targeting human host proteins that interact with SARS-CoV-2.

Specifications
Specifications
CAS | 1069-66-5 |
Molecular Formula | C8H15NaO2 |
Quantity | 100 mg |
Synonym | sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin |
InChI Key | AEQFSUDEHCCHBT-UHFFFAOYSA-M |
SMILES | CCCC(CCC)C(=O)[O-].[Na+] |
IUPAC Name | sodium;2-propylpentanoate |
Molecular Weight (g/mol) | 166.196 |
PubChem CID | 16760703 |
ChEBI | CHEBI:9925 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction