missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva™ Whatman™ Replacement Valve for MD110 Series Pressure Filtration Manifolds
Replacement Valve (model: MD110/03/10) for use with MD110 Series Pressure Filtration Manifolds.
Brand: Cytiva™ 10451014
This item is not returnable.
View return policy
Description
- Product Type: Valve
- Model: MD110/03/10
- For Use With (Equipment): MD110 Series Pressure Filtration Manifolds
- Quantity: 1 Pk.
Specifications
MD 110/03/10 | |
MD110 Series Pressure Filtration Manifolds | |
MD110/03/10 | |
1 Ea. | |
Valve |
Reusable | |
Non-sterile | |
1/Pk. | |
MD 110 |