Learn More
VAMP4 Mouse anti-Human, CoraLite 488, Clone: 3A6F2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67219-100UL
This item is not returnable.
View return policy
Description
VAMP4 is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. Characterized by a common sequence called the SNARE motif, SNARE proteins are involved in membrane fusion and vesicular transport (PMID: 11252968). VAMP4 may play a role in trans-Golgi network to endosome transport.
Specifications
VAMP4 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
IgG2a |
Immunofluorescence | |
3A6F2 | |
CoraLite 488 | |
100 μL | |
8674 | |
Liquid |