missing translation for 'onlineSavingsMsg'
Learn More

Vanadium carbide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11376508
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Vanadium carbide is used as an additive to tungsten carbide to fine the carbide crystals to improve the property of the cermets. It used in cutting tools and the steel industry. Used as material for research in electronics. Used anode materials for lithium ion batteries, and in supercapacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-10-9
Molecular Formula CV
Molecular Weight (g/mol) 62.95
MDL Number MFCD00049698
InChI Key ZLANVVMKMCTKMT-UHFFFAOYSA-N
IUPAC Name methanidylidynevanadiumylium
SMILES [C-]#[V+]

Specifications

Melting Point 2810°C
Color Black
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% metal basis
Solubility Information Insoluble in water.
Formula Weight 62.95
Odor Odorless
Chemical Name or Material Vanadium carbide
compliance-icons
Product Identifier
  • Vanadium carbide
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Vanadium carbide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'