missing translation for 'onlineSavingsMsg'
Learn More

Vanadium foil, 0.127mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11321319
Click to view available options
Quantity:
50 x 100 mm
100 x 200 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: approx. 3.87g/50x100mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]

Specifications

Color Silver to Gray
Appearance Silver Gray foil
Assay Percent Range 99.8% (Metals basis)
Quantity 100 x 200 mm
Odor Odorless
Form Foil
Chemical Name or Material Vanadium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'