missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) bromide, 99.5% (metals basis), Thermo Scientific Chemicals

76.08 € - 1231.13 €

Chemical Identifiers

CAS 13470-26-3
Molecular Formula Br3V
Molecular Weight (g/mol) 290.654
MDL Number MFCD00049695
InChI Key ZOYIPGHJSALYPY-UHFFFAOYSA-K
Synonym vanadium bromide, vanadium tribromide, vanadium bromide vbr3, vanadium iii bromide, wln: va e3, vanadium iii bromide-v
PubChem CID 83509
IUPAC Name tribromovanadium
SMILES [V](Br)(Br)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11334929
View Documents
Thermo Scientific Alfa Aesar
081135.04
2 g
76.08 €
2g
Please to purchase this item. Need a web account? Register with us today!
11344929
View Documents
Thermo Scientific Alfa Aesar
081135.09
10 g
286.43 €
10g
Please to purchase this item. Need a web account? Register with us today!
11324929
View Documents
Thermo Scientific Alfa Aesar
081135.18
50 g
1231.13 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vanadium(III) bromide is used as a catalyst for C(sp3)-H fluorination reactions especially the benzylic fluorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13470-26-3
290.654
ZOYIPGHJSALYPY-UHFFFAOYSA-K
83509
[V](Br)(Br)Br
Br3V
MFCD00049695
vanadium bromide, vanadium tribromide, vanadium bromide vbr3, vanadium iii bromide, wln: va e3, vanadium iii bromide-v
tribromovanadium

Specifications

(decomposition)
Crystalline
Br3V
vanadium bromide, vanadium tribromide, vanadium bromide vbr3, vanadium iii bromide, wln: va e3, vanadium iii bromide-v
ZOYIPGHJSALYPY-UHFFFAOYSA-K
tribromovanadium
83509
Moisture sensitive
Vanadium(III) bromide
13470-26-3
99.5% (metals basis)
MFCD00049695
Soluble in water and tetrahydrofuran.
[V](Br)(Br)Br
290.654
290.67
4.00^1^8 g/mL
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 236-736-6

missing translation for 'rtecsNumber' : YW2750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.