missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) chloride, 99% (metals basis), Thermo Scientific Chemicals

83.82 € - 413.06 €

Chemical Identifiers

CAS 7718-98-1
Molecular Formula Cl3V
Molecular Weight (g/mol) 157.29
MDL Number MFCD00011454
InChI Key HQYCOEXWFMFWLR-UHFFFAOYSA-K
PubChem CID 62647
IUPAC Name trichlorovanadium
SMILES [Cl-].[Cl-].[Cl-].[V+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11321579
SDS View Documents
Thermo Scientific Alfa Aesar
014039.09
10 g
83.82 €
10g
Please to purchase this item. Need a web account? Register with us today!
11311579
SDS View Documents
Thermo Scientific Alfa Aesar
014039.18
50 g
413.06 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vanadium(III) chloride acts as an intermediate in organic synthesis and in the manufacture of organic light emitting diodes. It acts as a precursor to other vanadium(III) complexes. Further, it is used in the preparation of vanadium dichloride and organovanadium compounds. In addition to this, it serves as a catalyst in the production of ethylene propylene diene monomer rubber and polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 231-744-6

missing translation for 'rtecsNumber' : YW2800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium(III) chloride, 99% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.