missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chl, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Red |
Physical Form | Solution |
Quantity | 800 mL |
Linear Formula | VCl·3THF |
Flash Point | −8°C |
Formula Weight | 373.62 |
Concentration or Composition (by Analyte or Components) | 0.40 to 0.60M |
Packaging | AcroSeal glass Bottle |
Specific Gravity | 1.312 |
Density | 1.3120g/mL |
Show More |
Safety and Handling




Product Identifier
- Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chloride
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction