Learn More
Vanadium(III) fluoride, 98%, Thermo Scientific Chemicals
138.69 € - 3366.75 €
Chemical Identifiers
CAS | 10049-12-4 |
---|---|
Molecular Formula | F3V |
Molecular Weight (g/mol) | 107.94 |
MDL Number | MFCD00042553 |
InChI Key | PASVSMBJEYQYSN-UHFFFAOYSA-K |
Synonym | Vanadium trifluoride |
IUPAC Name | vanadium(3+) trifluoride |
SMILES | [F-].[F-].[F-].[V+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314919
|
Thermo Scientific Alfa Aesar
081115.04 |
2 g |
138.69 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324919
|
Thermo Scientific Alfa Aesar
081115.09 |
10 g |
678.38 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334919
|
Thermo Scientific Alfa Aesar
081115.18 |
50 g |
3366.75 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Employed in oxygen-sensitive applications, such as metal production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10049-12-4 | |
107.94 | |
PASVSMBJEYQYSN-UHFFFAOYSA-K | |
vanadium(3+) trifluoride |
F3V | |
MFCD00042553 | |
Vanadium trifluoride | |
[F-].[F-].[F-].[V+3] |
Specifications
≥1406°C | |
Powder | |
MFCD00042553 | |
14,9922 | |
Practically insoluble in water, acetone, and glacial acetic acid | |
[F-].[F-].[F-].[V+3] | |
107.94 | |
98% | |
3.363 g/mL at 19°C |
10049-12-4 | |
F3V | |
UN2923 | |
Vanadium trifluoride | |
PASVSMBJEYQYSN-UHFFFAOYSA-K | |
vanadium(3+) trifluoride | |
107.94 | |
Moisture sensitive | |
Vanadium (III) fluoride |
Safety and Handling
GHS H Statement
H300-H330-H314-H318
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H330-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 233-169-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only