Learn More
Vanadium(III) oxide, 95%, Thermo Scientific Chemicals
Description
Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1970°C |
Color | Black |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 95% |
Solubility Information | Insoluble in water |
IUPAC Name | oxo[(oxovanadio)oxy]vanadium |
Formula Weight | 149.88 |
Odor | Odorless |
Density | 4.87 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M13L011 | Certificate of Analysis | 15/01/2025 | 012301.30,012301.A1,012301.A7 |
X25J024 | Certificate of Analysis | 07/11/2023 | 012301.30,012301.A1,012301.A7 |
T17J017 | Certificate of Analysis | 26/07/2023 | 012301.30,012301.A1,012301.A7 |
T22F029 | Certificate of Analysis | 25/10/2022 | 012301.30,012301.A1,012301.A7 |
H14S021 | Certificate of Analysis | 08/12/2021 | 012301.30,012301.A1,012301.A7 |
Safety and Handling

- Vanadium(III) oxide
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Vanadium oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.