Learn More
Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
94.37 € - 374.87 €
Chemical Identifiers
CAS | 12036-21-4 |
---|---|
Molecular Formula | O2V |
Molecular Weight (g/mol) | 82.94 |
MDL Number | MFCD00049702 |
InChI Key | GRUMUEUJTSXQOI-UHFFFAOYSA-N |
Synonym | vanadium dioxide, vanadium iv oxide, vanadium oxide vo2, vo2, vanadium iv oxide trace metals basis 10g |
PubChem CID | 82849 |
ChEBI | CHEBI:30047 |
IUPAC Name | dioxovanadium |
SMILES | O=[V]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308237
|
Thermo Scientific Alfa Aesar
022957.09 |
10 g |
94.37 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398227
|
Thermo Scientific Alfa Aesar
022957.18 |
50 g |
374.87 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vanadium(IV) oxide is used in chemical sensors, energy-conserving coatings, transparent conductors and switching materials. It acts as a stationary optical shutter, optical modulators, cameras and data storage. As infrared modulators, it is used for the missile guidance systems. Thin films made up of vanadium dioxide find applications in electro-optical switches, micro-optical switch, passive smart radiators and sunshields for spacecraft.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12036-21-4 | |
82.94 | |
GRUMUEUJTSXQOI-UHFFFAOYSA-N | |
82849 | |
dioxovanadium |
O2V | |
MFCD00049702 | |
vanadium dioxide, vanadium iv oxide, vanadium oxide vo2, vo2, vanadium iv oxide trace metals basis 10g | |
CHEBI:30047 | |
O=[V]=O |
Specifications
1967°C | |
Powder | |
O2V | |
vanadium dioxide, vanadium iv oxide, vanadium oxide vo2, vo2, vanadium iv oxide trace metals basis 10g | |
GRUMUEUJTSXQOI-UHFFFAOYSA-N | |
dioxovanadium | |
82849 | |
82.94 | |
Vanadium(IV) oxide |
12036-21-4 | |
99% (metals basis) | |
MFCD00049702 | |
Insoluble in water. | |
O=[V]=O | |
82.94 | |
CHEBI:30047 | |
4.339 g/mL |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 234-841-1
missing translation for 'tsca' : Yes
- Vanadium(IV) oxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Vanadium oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.