missing translation for 'onlineSavingsMsg'
Learn More
Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals Product Code.: 11374919

Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

Product Code. 11374919
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11374919

Brand: Thermo Scientific Alfa Aesar 081119.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment

Employed as a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment. It is used in organic chemistry as a reagent in the epoxidation of allylic alcohols in combination with tert-butyl hydroperoxide (TBHP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-26-2
Molecular Formula C10H14O5V
Molecular Weight (g/mol) 265.16
MDL Number MFCD00000032
InChI Key JFHJZWAQYMGNBE-SUKNRPLKSA-L
Synonym vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
PubChem CID 131674261
IUPAC Name (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
SMILES O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Boiling Point 140°C (13mmHg)
Melting Point 256°C to 259°C
Physical Form Powder
Quantity 10 g
UN Number UN3285
Solubility Information Moderately soluble in acetone, ether and chloroform; Soluble in ethanol and benzene
Formula Weight 265.16
Density 1.4 g/mL
Chemical Name or Material Vanadium (IV) oxide bis (2,4-pentanedionate)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.