missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) oxide meso-tetraphenylporphine, Thermo Scientific Chemicals

52.06 € - 177.89 €

Chemical Identifiers

CAS 14705-63-6
Molecular Formula C44H30N4O2V
Molecular Weight (g/mol) 697.692
MDL Number MFCD00012154
InChI Key LRQXAIDMLNPKOR-UHFFFAOYSA-N
Synonym 5,10,15,20-tetraphenyl-21h,23h-porphine vanadium iv oxide
PubChem CID 131699123
IUPAC Name oxygen(2-);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;vanadium(4+)
SMILES C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[O-2].[O-2].[V+4]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11460231
SDS View Documents
Thermo Scientific Alfa Aesar
039583.MC
100 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11314497
SDS View Documents
Thermo Scientific Alfa Aesar
039583.ME
500 mg
177.89 €
500mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used for research in catalysis. Thin films of VOTPP may be used in detecting methane gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14705-63-6
697.692
LRQXAIDMLNPKOR-UHFFFAOYSA-N
131699123
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[O-2].[O-2].[V+4]
C44H30N4O2V
MFCD00012154
5,10,15,20-tetraphenyl-21h,23h-porphine vanadium iv oxide
oxygen(2-);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;vanadium(4+)

Specifications

14705-63-6
Crystalline
MFCD00012154
Insoluble in water.
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[O-2].[O-2].[V+4]
697.692
679.67
Vanadium(IV) oxide meso-tetraphenylporphine
Purple
C44H30N4O2V
5,10,15,20-tetraphenyl-21h,23h-porphine vanadium iv oxide
LRQXAIDMLNPKOR-UHFFFAOYSA-N
oxygen(2-);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;vanadium(4+)
131699123
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.