missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium Pentoxide, Pure, Fisher Chemical™
67.30 € - 228.00 €
Chemical Identifiers
CAS | 1314-62-1 |
---|---|
Molecular Formula | O5V2 |
Molecular Weight (g/mol) | 181.88 |
MDL Number | MFCD00011457 |
InChI Key | GNTDGMZSJNCJKK-UHFFFAOYSA-N |
Synonym | vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride |
PubChem CID | 14814 |
ChEBI | CHEBI:30045 |
IUPAC Name | dioxovanadiooxy(dioxo)vanadium |
SMILES | O=[V](=O)O[V](=O)=O |
Description
Chemical Identifiers
1314-62-1 | |
181.88 | |
GNTDGMZSJNCJKK-UHFFFAOYSA-N | |
14814 | |
dioxovanadiooxy(dioxo)vanadium |
O5V2 | |
MFCD00011457 | |
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride | |
CHEBI:30045 | |
O=[V](=O)O[V](=O)=O |
Specifications
1750°C | |
1314-62-1 | |
4 | |
O5V2 | |
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride | |
O=[V](=O)O[V](=O)=O | |
181.88 | |
CHEBI:30045 | |
Plastic powder jar |
690°C | |
Amber | |
Powder/Solid | |
MFCD00011457 | |
GNTDGMZSJNCJKK-UHFFFAOYSA-N | |
dioxovanadiooxy(dioxo)vanadium | |
14814 | |
181.88g/mol | |
0.0443 hPa 700 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Vanadium(V) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.