missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7440-62-2 |
---|---|
Molecular Formula | V |
Molecular Weight (g/mol) | 50.94 |
MDL Number | MFCD00011453 |
InChI Key | LEONUFNNVUYDNQ-UHFFFAOYSA-N |
Synonym | dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde |
PubChem CID | 23990 |
ChEBI | CHEBI:35170 |
IUPAC Name | vanadium |
SMILES | [V] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311519
|
Thermo Scientific Alfa Aesar
013996.BQ |
5 cm |
N/A
|
N/A | |||||
11301519
|
Thermo Scientific Alfa Aesar
013996.BU |
25 cm |
N/A
|
N/A | |||||
11391509
|
Thermo Scientific Alfa Aesar
013996.BY |
100 cm |
N/A
|
N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-62-2 | |
50.94 | |
LEONUFNNVUYDNQ-UHFFFAOYSA-N | |
23990 | |
vanadium |
V | |
MFCD00011453 | |
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde | |
CHEBI:35170 | |
[V] |
Specifications
7440-62-2 | |
MFCD00011453 | |
LEONUFNNVUYDNQ-UHFFFAOYSA-N | |
vanadium | |
23990 | |
99.5% | |
≈1.93g/cm | |
(metals basis) |
V | |
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde | |
[V] | |
50.94 | |
CHEBI:35170 | |
Odorless | |
Rod | |
Vanadium rod, 6.35mm (0.25 in.) dia. |
Safety and Handling
missing translation for 'einecsNumber' : 231-171-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only