Learn More
Vanadium(V) oxide, 99.2%, Thermo Scientific Chemicals
53.67 € - 182.91 €
Chemical Identifiers
CAS | 1314-62-1 |
---|---|
Molecular Formula | O5V2 |
Molecular Weight (g/mol) | 181.88 |
MDL Number | MFCD00011457 |
InChI Key | GNTDGMZSJNCJKK-UHFFFAOYSA-N |
Synonym | vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride |
PubChem CID | 14814 |
ChEBI | CHEBI:30045 |
SMILES | O=[V](=O)O[V](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386108
|
Thermo Scientific Alfa Aesar
011093.14 |
25 g |
53.67 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376108
|
Thermo Scientific Alfa Aesar
011093.30 |
250 g |
75.38 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366108
|
Thermo Scientific Alfa Aesar
011093.A1 |
1 kg |
182.91 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
As an oxidation catalyst, in refractories, as a ceramics colorant, in the manufacture of vanadium salts, and as a dye.Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst in the manufacturing of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms. Additionally, it is used as ceramics colorant, as dye and also as a detector material in bolometers in the field of thermal imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- As an oxidation catalyst, in refractories, as a ceramics colorant, in the manufacture of vanadium salts, and as a dye
Chemical Identifiers
1314-62-1 | |
181.88 | |
GNTDGMZSJNCJKK-UHFFFAOYSA-N | |
14814 | |
O=[V](=O)O[V](=O)=O |
O5V2 | |
MFCD00011457 | |
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride | |
CHEBI:30045 |
Specifications
1750°C (decomposition) | |
1314-62-1 | |
O5V2 | |
UN2862 | |
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride | |
GNTDGMZSJNCJKK-UHFFFAOYSA-N | |
dioxovanadiooxy(dioxo)vanadium | |
14814 | |
181.88 | |
Odorless | |
Vanadium(V) oxide |
690°C | |
Powder | |
MFCD00011457 | |
14,9921 | |
Slightly soluble in water; Soluble in acids and alkalies. | |
O=[V](=O)O[V](=O)=O | |
181.88 | |
CHEBI:30045 | |
99.2% | |
3.357 g/mL |
Safety and Handling
GHS H Statement
H372-H341-H361-H302-H332-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P501c
H302+H332-H318-H335-H341-H351-H361d-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM PENTOXIDE
missing translation for 'einecsNumber' : 215-239-8
missing translation for 'rtecsNumber' : YW2125000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Vanadium(V) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- Reproductive toxicity on or via lactation
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H362-May cause harm to breast-fed children.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Vanadium oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.