missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) oxide, 99.6% min, Thermo Scientific Chemicals

46.03 € - 190.95 €

Chemical Identifiers

CAS 1314-62-1
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
Synonym vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
PubChem CID 14814
ChEBI CHEBI:30045
IUPAC Name dioxovanadiooxy(dioxo)vanadium
SMILES O=[V](=O)O[V](=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11306118
SDS View Documents
Thermo Scientific Alfa Aesar
011094.09
10 g
46.03 €
10g
Please to purchase this item. Need a web account? Register with us today!
11396108
SDS View Documents
Thermo Scientific Alfa Aesar
011094.22
100 g
57.39 €
100g
Please to purchase this item. Need a web account? Register with us today!
11316118
SDS View Documents
Thermo Scientific Alfa Aesar
011094.36
500 g
190.95 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst for the manufacture of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms. Additionally, it is used as ceramics colorant, as dye and also as a detector material in bolometers in the field of thermal imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-62-1
181.88
GNTDGMZSJNCJKK-UHFFFAOYSA-N
14814
dioxovanadiooxy(dioxo)vanadium
O5V2
MFCD00011457
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
CHEBI:30045
O=[V](=O)O[V](=O)=O

Specifications

1750°C (decomposition)
1314-62-1
O5V2
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
GNTDGMZSJNCJKK-UHFFFAOYSA-N
dioxovanadiooxy(dioxo)vanadium
14814
181.88
Vanadium(V) oxide
690°C
99.6% min
MFCD00011457
Slightly soluble in water.
O=[V](=O)O[V](=O)=O
181.88
CHEBI:30045
3.357 g/mL
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P501c

H302+H332-H318-H335-H341-H351-H361d-H372

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 215-239-8

missing translation for 'rtecsNumber' : YW2125000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.