missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Vacuum deposition

327.00 € - 948.72 €

Chemical Identifiers

CAS 1314-62-1
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
Synonym vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
PubChem CID 14814
ChEBI CHEBI:30045
IUPAC Name dioxovanadiooxy(dioxo)vanadium
SMILES O=[V](=O)O[V](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11460710
View Documents
Thermo Scientific Alfa Aesar
044424.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11470710
View Documents
Thermo Scientific Alfa Aesar
044424.22
100 g
948.72 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vanadium(V) oxide is used in Vacuum deposition, catalysts for conversion of toluene to benzonitrile or propylene to acrylonitrile, as a detector material in bolometers and microbolometer arrays for thermal imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-62-1
181.88
GNTDGMZSJNCJKK-UHFFFAOYSA-N
14814
dioxovanadiooxy(dioxo)vanadium
O5V2
MFCD00011457
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
CHEBI:30045
O=[V](=O)O[V](=O)=O

Specifications

1750°C (decomposition)
1314-62-1
O5V2
UN2862
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
GNTDGMZSJNCJKK-UHFFFAOYSA-N
dioxovanadiooxy(dioxo)vanadium
14814
181.88
Odorless
(metals basis)
690°C
Pieces
MFCD00011457
14,9921
Soluble in water(8g/L).
O=[V](=O)O[V](=O)=O
181.88
CHEBI:30045
99.9%
3.357 g/mL
Vanadium (V) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H341-H361-H302-H332-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P501c

H302+H332-H318-H335-H341-H351-H361d-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM PENTOXIDE

missing translation for 'einecsNumber' : 215-239-8

missing translation for 'rtecsNumber' : YW2125000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only