missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) trichloride oxide, V+5 28.5% min

Product Code. p-7080313
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 11354919

Brand: Thermo Scientific Alfa Aesar 081118.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7727-18-6 | Cl3OV | 173.29 g/mol

Vanadium(V) trichloride oxide is used as a catalyst in olefin polymerization reaction, especially for the production of ethylene-propylene rubber. It is also used to synthesis organovanadium compounds and as a mordant in dyeing. It acts as a precursor to prepare vanadium pentoxide, vanadyl dichloride and dioxovanadium monochloride. Further, it serves as a reagent and a strong oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7727-18-6
Molecular Formula Cl3OV
Molecular Weight (g/mol) 173.29
MDL Number MFCD00011459
InChI Key WMZFXRASJAIALG-UHFFFAOYSA-K
Synonym vanadyl trichloride, trichlorooxovanadium, vanadium v trichloride oxide, trichlorooxovanadium oxide, vanadium v oxytrichloride, trichlorooxo vanadium, vanadium, trichlorooxo-, t-4, vanadium, trichlorooxo, vanadium oxide trichloride, vanadium trichloride oxide
PubChem CID 24410
IUPAC Name oxovanadium;trihydrochloride
SMILES [Cl-].[Cl-].[Cl-].O=[V+5]

Spezifikation

Boiling Point 126°C to 127°C
Melting Point -77°C
Color Yellow
Physical Form Liquid
Quantity 10 g
UN Number UN2443
Merck Index 14,9927
Solubility Information Miscible with benzene,dichloromethane and hexane.
Formula Weight 173.3
Concentration or Composition (by Analyte or Components) V+5 28.5% min
Odor Chlorine-like
Sensitivity Moisture sensitive
Density 1.829 g/mL
Chemical Name or Material Vanadium (V) trichloride oxide
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Vanadium(V) trichloride oxide
Signalwort
  • Danger
Gefahrenkategorie
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Gefahrenhinweis
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Vanadium oxytrichloride

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt