missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) triisopropoxide oxide, 96%, Thermo Scientific Chemicals

Product Code. 11423830
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Vanadium(V) triisopropoxide oxide serves as a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands. It is also used in catalysis. Further, it acts as a precursor and used in thermal atomic layer deposition of vanadium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5588-84-1
Molecular Formula C9H24O4V
Molecular Weight (g/mol) 247.23
MDL Number MFCD00015017
InChI Key JOUSPCDMLWUHSO-UHFFFAOYSA-N
Synonym vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n
PubChem CID 79702
IUPAC Name oxovanadium;propan-2-ol
SMILES O=[V].CC(C)O.CC(C)O.CC(C)O

Specifications

Boiling Point 81°C to 83°C (2mmHg)
Melting Point -12°C
Color Orange-Yellow
Physical Form Liquid
Quantity 500 g
Linear Formula VO[CHO(CH3)2]3
UN Number UN1993
Flash Point 45°C (113°F)
Solubility Information Miscible with toluene,hexane and isopropanol.
Formula Weight 242.2
Percent Purity 96%
Sensitivity Moisture sensitive
Density 1.04 g/mL
Chemical Name or Material Vanadium (V) triisopropoxide oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'