missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) triisopropoxide oxide, 96%, Thermo Scientific Chemicals

Product Code. 11443830
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Vanadium(V) triisopropoxide oxide serves as a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands. It is also used in catalysis. Further, it acts as a precursor and used in thermal atomic layer deposition of vanadium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5588-84-1
Molecular Formula C9H24O4V
Molecular Weight (g/mol) 247.23
MDL Number MFCD00015017
InChI Key JOUSPCDMLWUHSO-UHFFFAOYSA-N
Synonym vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n
PubChem CID 79702
IUPAC Name oxovanadium;propan-2-ol
SMILES O=[V].CC(C)O.CC(C)O.CC(C)O

Specifications

Boiling Point 81°C to 83°C (2mmHg)
Melting Point -12°C
Color Orange-Yellow
Physical Form Liquid
Quantity 1 g
Linear Formula VO[CHO(CH3)2]3
UN Number UN1993
Flash Point 45°C (113°F)
Solubility Information Miscible with toluene,hexane and isopropanol.
Formula Weight 242.2
Percent Purity 96%
Sensitivity Moisture sensitive
Density 1.04 g/mL
Chemical Name or Material Vanadium (V) triisopropoxide oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Vanadium(V) triisopropoxide oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Vanadium(V) triisopropoxide oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.