Learn More
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
45.80 € - 440.00 €
Chemical Identifiers
CAS | 3153-26-2 |
---|---|
Molecular Formula | C10H14O5V |
Molecular Weight (g/mol) | 265.16 |
MDL Number | MFCD00000032 |
InChI Key | JFHJZWAQYMGNBE-SUKNRPLKSA-L |
Synonym | vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide |
PubChem CID | 131674261 |
IUPAC Name | (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate |
SMILES | O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10163880
|
Thermo Scientific Acros
199590500 |
50 g | Glass bottle |
45.80 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10338340
|
Thermo Scientific
199592500 |
250 g | Glass bottle |
149.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10444795
|
Thermo Scientific Acros
199590010 |
1 kg | Glass bottle |
440.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3153-26-2 | |
265.16 | |
JFHJZWAQYMGNBE-SUKNRPLKSA-L | |
131674261 | |
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
C10H14O5V | |
MFCD00000032 | |
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide | |
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate |
Specifications
3153-26-2 | |
Blue-Green to Green | |
Authentic | |
C10H14O5V | |
MFCD00000032 | |
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide | |
JFHJZWAQYMGNBE-SUKNRPLKSA-L | |
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate | |
131674261 | |
99% | |
Vanadyl(IV) acetylacetonate, 99% |
256.0°C to 259.0°C | |
79°C | |
98.5% min | |
(CH3COCH=C(CH3)O)2V=O | |
02,456; 03,331; 11,47; 13,54; 15,57; 15,186 | |
Solubility in water: negligible. Other solubilities: soluble in methanol (6.4 g/100 mL), ethanol, ether, chloroform, acetone, dioxane, benzene (0.9 g/100 m, l), pyridine | |
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
265.16 | |
265.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If eye irritation persists: Get medical
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 221-590-8
missing translation for 'tsca' : TSCA
- Vanadyl(IV) acetylacetonate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only