Learn More
Testo™ Vane Anemometer with Humidity Measurement
Brand:  Testo™ 0560 4102
Product Code. 10233183
  • 341.00 € / Each
Explore more special offers
Specifications
Vane Anemometer
133 x 46 x 25 mm
-20 to +70°C
±0.5 m/sec.
Measures air flow, air temperature, and humidity