Learn More
ATP™ Vane Velocity Digital Thermo-Anemometer
Brand:  ATP™ AVM-618B
Encompass
Product Code. 12705095
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Vane Velocity Thermoanemometer
150 x 72 x 35 mm
0°C to 50°C
±3%
9V PP3 Alkaline Battery
350 g