missing translation for 'onlineSavingsMsg'
Learn More

Vanillin, 99%, pure

Product Code. 11984781 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
100 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
250g
2g
5kg
This item is not returnable. View return policy

Product Code. 11984781

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-33-5 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-33-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00006942,MFCD08702848
InChI Key MWOOGOJBHIARFG-UHFFFAOYSA-N
Synonym vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
PubChem CID 1183
ChEBI CHEBI:18346
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1O

Specifications

Melting Point 81.0°C to 84.0°C
Color White to Yellow
Density 1.0600g/mL
Boiling Point 170.0°C (15.0 mmHg)
Flash Point 147°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic drum
Quantity 5 kg
Beilstein 08, 247
Merck Index 15, 10120
Solubility Information Solubility in water: 10g/L (25°C). Other solubilities: freely soluble in glacial acetic acid,freely soluble in alcohol,chloroform,ether,,carbon disulfide and methanol,soluble in oils,aq. solutions of alkali hydroxide,s
Formula Weight 152.15
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Vanillin
Show More Show Less
compliance-icons
Product Identifier
  • Vanillin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.